Få mere at vide
Tocris Bioscience™ AMG PERK 44
GREENER_CHOICE
Potent and selective PERK inhibitor; orally bioavailable
Brand:  Tocris Bioscience™ 5517/10
Encompass
Produktkode 15786725
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
AMG PERK 44 is a potent and selective PERK inhibitor (IC50 = 6 nM). Exhibits >160-fold selectivity for PERK over B-Raf and GCN2, as well as a panel of 387 other kinases. Orally bioavailable.
Tekniske data
Tekniske data
4-[2-Amino-4-methyl-3-(2-methyl-6-quinolinyl)benzoyl]-1,2-dihydro-1-methyl-2,5-diphenyl-3H-pyrazol-3-one hydrochloride
1883548-84-2
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner