Få mere at vide
Tocris Bioscience™ AMG 837 hemicalcium salt
Potent free fatty acid receptor 1 (FFA1/GPR40) partial agonist
Brand: Tocris Bioscience™ 6082/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
AMG 837 hemicalcium salt is a potent free fatty acid receptor 1 (FFA1/GPR40) partial agonist (EC50 values are 13.5, 22.6 and 31.7 nM at human, mouse and rat receptors, respectively). Interacts allosterically with full FFA1 agonist Docosahexaenoic acid (Cat. No. 3687). Potentiates glucose-dependent stimulation of insulin in vivo. Antidiabetic.Tekniske data
| AMG 837 hemicalcium salt | |
| (βS)-β-1-Propyn-1-yl-4-[[4'-(trifluoromethyl)[1,1'-biphenyl]-3-yl]methoxy]benzenepropanoic acid hemicalcium salt | |
| Soluble to 100mM in DMSO | |
| White |
| 1291087-14-3 | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only.