Få mere at vide
Tocris Bioscience™ AMG 21629
Potent and selective TRPV1 antagonist
Brand: Tocris Bioscience™ 4330/1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
AMG 21629 is a potent and selective TRPV1 antagonist. Blocks Ca2+ uptake by CHO cells expressing TRPV1 receptors (IC50 values are 0.6 and 0.8 nM for capsaicin (Cat. No. 0462) and acid-induced Ca2+ uptake, respectively). Exhibits >4000-fold selectivity for TRPV1 over other TRP channels. Blocks capsaicin-induced flinch response and causes hyperthermia in rats. Orally available and non-CNS penetrant.Tekniske data
| 3-Amino-5-[[2-[(2-methoxyethyl)amino]-6-[4-(trifluoromethyl)phenyl]-4-pyrimidinyl]oxy]-2(1H)-quinoxalinone | |
| 939040-79-6 | |
| 1 mg |