Få mere at vide
AMG 18 hydrochloride, Tocris Biosciences™
IRE1 Inhibitors
Brand:  Tocris Bioscience 6166/5
Encompass
Produktkode 15997910
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
AMG 18 hydrochloride is a potent and selective IRE1α inhibitor (IC50 values are 13 and 99 nM for recombinant and cellular IRE1α, respectively). Exhibits selectivity for IRE1α in a panel of over 100 other kinases.
Tekniske data
Tekniske data
2250019-92-0
2-Chloro-N-[6-methyl-5-[[3-[2-[(3S)-3-piperidinylamino]-4-pyrimidinyl]-2-pyridinyl]oxy]-1-naphthalenyl]benzenesulfonamide hydrochloride
5 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater