Få mere at vide
Ametryn, TRC
Artikelnummer. 17977599
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17977599 5 g
17957599 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17977599 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5764005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 834-12-8
Molekylær formel C9 H17 N5 S
Molekylvægt (g/mol) 227.33
Synonym 1,3,5-Triazine-2,4-diamine, N-ethyl-N'-(1-methylethyl)-6-(methylthio)- (9CI), s-Triazine, 2-(ethylamino)-4-(isopropylamino)-6-(methylthio)- (6CI,8CI), N2-Ethyl-N4-(1-methylethyl)-6-(methylthio)-1,3,5-triazine-2,4-diamine, 2-Ethylamino-4-isopropylamino-6-methylmercapto-s-triazine, 2-Ethylamino-4-isopropylamino-6-methylthio-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-methylthio-s-triazine, 2-Ethylamino-4-methylthio-6-isopropylamino-1,3,5-triazine, 2-Methylmercapto-4-ethylamino-6-isopropylamino-s-triazine, 2-Methylthio-4-ethylamino-6-isopropylamino-s-triazine, 2-Methylthio-4-ethylamino-6-isopropylamino-sym-triazine, A 1093, Amephyt, Ametrex, Ametryn, Ametryne, Cemerin, Doruplant, Evik, G 34162, Gesapax, Metrimex 500SC, N-Ethyl-N'-(1-methylethyl)-6-(methylthio)-1,3,5-triazine-2,4-diamine, NSC 163044, Primatol Z 80
IUPAC navn 4-N-ethyl-6-methylsulfanyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
SMIL CCNc1nc(NC(C)C)nc(SC)n1
Tekniske data
Kemisk navn eller materiale Ametryn
Mængde 5 g
InChI formel InChI=1S/C9H17N5S/c1-5-10-7-12-8(11-6(2)3)14-9(13-7)15-4/h6H,5H2,1-4H3,(H2,10,11,12,13,14)
Formel vægt 227.1205