missing translation for 'onlineSavingsMsg'
Få mere at vide

Amenamevir, MedChemExpress

Artikelnummer. 16466556
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16466556 100 mg
16436188 10 mg
16408787 5 mg
16402327 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16466556 Leverandør Medchem Express Leverandørnr. HY14809

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Amenamevir is a helicase-primase inhibitor which has potent antiviral activity against HSVs with an EC50 of 14 ng/mL.

Amenamevir is a helicase-primase inhibitor which has potent antiviral activity against HSVs with an EC50 of 14 ng/mL.

Specifications

Kemisk navn eller materiale Amenamevir
Fysisk form Solid
CAS 841301-32-4
Farve Light Yellow
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C24H26N4O5S
Til brug med (applikation) COVID-19-anti-virus
Synonym ASP2151
Opløselighedsinformation DMSO : ≥ 50 mg/mL (103.62 mM)
SMIL O=C(C(CC1)CCS1(=O)=O)N(C2=C(C)C=CC=C2C)CC(NC3=CC=C(C4=NOC=N4)C=C3)=O
Molekylvægt (g/mol) 482.55
Formel vægt 482.55
Procent renhed 99.89%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.