Få mere at vide
Amdinocillin, TRC
Artikelnummer. 30138037
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138037 1 g
30135297 500 mg
17948134 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138037 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5763001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32887-01-7
Molekylær formel C15 H23 N3 O3 S
Molekylvægt (g/mol) 325.43
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (+)- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, [2S-(2α,5α,6β)]-, (2S,5R,6R)-6-[[(Hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(Hexahydro-1H-azepin-1-yl)methylene]amino]penicillanic acid, 6β-[(Hexahydro-1H-azepin-1-yl)methyleneamino]penicillanic acid, Amdinocillin, Coactin, FL 1060, Hexacillin, Hexapen, Mecillinam, Penicillin Hx
IUPAC navn (2S,5R,6R)-6-[(E)-azepan-1-ylmethylideneamino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMIL CC1(C)S[C@@H]2[C@H](\N=C\N3CCCCCC3)C(=O)N2[C@H]1C(=O)O
Tekniske data
Kemisk navn eller materiale Mecillinam
Analyt- eller komponentnavne Mecillinam
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C15H23N3O3S/c1-15(2)11(14(20)21)18-12(19)10(13(18)22-15)16-9-17-7-5-3-4-6-8-17/h9-11,13H,3-8H2,1-2H3,(H,20,21)/b16-9+/t10-,11+,13-/m1/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 1 g
Formel vægt 325.146
Procent renhed >95%