Få mere at vide
Tocris Bioscience™ AMD 3100 octahydrochloride
GREENER_CHOICE
Artikelnummer. 30001448
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001448 10 mg
30001494 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001448 Leverandør Tocris Bioscience™ Leverandørnr. 3299/10
Highly selective CXCR4 antagonist
Highly selective CXCR4 chemokine receptor antagonist (IC50 values are 0.02 - 0.13 and > 25 μM for CXCR4 and all other chemokine receptors respectively). Switches inflammatory responses from Th2 to Th1 type and reduces airway hyperresponsiveness in a mouse model of asthma. Potently inhibits HIV-1 and HIV-2 replication in vitro (EC50 = 4 - 35 nM) and mobilizes hematopoietic stem cells in vivo. Attenuates cocaine place preference and locomotor stimulation in rats. Attenuates microglial activation neurological function after ischemic stroke in mice.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 155148-31-5
Molekylær formel C28H62Cl8N8
Tekniske data
Kemisk navn eller materiale AMD 3100 octahydrochloride
Mængde 10 mg
Formel vægt 794.48
Renhed >98%
Molekylvægt (g/mol) 794.46