missing translation for 'onlineSavingsMsg'
Få mere at vide

Ambroxol O-glucuronide, TRC

Artikelnummer. 30322625
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322625 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30322625 Leverandør Toronto Research Chemicals Leverandørnr. TRCA57592550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Ambroxol O-beta-glucuronide
CAS 1241045-91-9
Molekylær formel C19H26Br2N2O7
Mængde 50 mg
Synonym trans-4-[[(2-Amino-3,5-dibromophenyl)methyl]amino]cyclohexylβ-D-glucopyranosiduronic Acid
InChI formel InChI=1S/C19H26Br2N2O7/c20-9-5-8(13(22)12(21)6-9)7-23-10-1-3-11(4-2-10)29-19-16(26)14(24)15(25)17(30-19)18(27)28/h5-6,10-11,14-17,19,23-26H,1-4,7,22H2,(H,27,28)/t10?,11?,14-,15-,16+,17-,19+/m0/s1
SMIL NC1=C(Br)C=C(Br)C=C1CN[C@@H]2CC[C@@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O3)CC2
IUPAC navn (2S,3S,4S,5R,6R)-6-[4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvægt (g/mol) 554.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.