Få mere at vide
Invitrogen™ Ambion™ Magna-Sep™ Magnetic Particle Separator
Mængde:
1 stk.
Pakningsstørrelse:
Hver
Beskrivelse
- The Magna-Sep Magnetic Particle Separator is available in three formats: for use with 1.5mL tubes, a six-tube format for use with pipettors, and a 96-well format for use with 12-channel pipettors
DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, RNA Extraction, RNA Isolation for Expression Microarrays, Transcriptome RNA Isolation, mRNA Isolation
Ordreinfo
Shipping Condition: Room Temperature
Tekniske data
Tekniske data
| Mængde | 1 stk. |