Learn More
Amberlite™ IRA-900 Cl ion-exchange resin, Thermo Scientific Chemicals
Catalog Number p-2885556
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
9050-97-9
C4H12ClN
109.60
MFCD00132712
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
6379
CHEBI:7070
[Cl-].C[N+](C)(C)C
9050-97-9
C4H12ClN
109.60
MFCD00132712
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
6379
CHEBI:7070
[Cl-].C[N+](C)(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
TRUSTED_SUSTAINABILITY
Physical Form Beads
Particle Size Particle size analysis : uniformity coefficient: <1.8, harmonic mean size: 0.650 to 0.820mm, <0.300mm: <0.5%,> 1.180mm: <3.0%
Packaging Plastic bottle
Volume Total Capacity (Cl-):> 1.0 eq/L
Water Retention Capacity (Cl-): 58 to 64%
Fieser 01,511; 02,267; 04,267; 06,302; 07,182
Color Beige/Brown to Ivory
Quantity 100 g
IUPAC Name tetramethylazanium;chloride
Chemical Name or Material Amberlite™ IRA-900