Learn More
Amberlite™ IRA-410, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
Specifications
Packaging | Plastic bottle |
Fieser | 01,511; 02,267; 04,267; 05,432; 06,302; 07,182 |
Quantity | 100 g |
Chemical Name or Material | Amberlite™ IRA-410 |