Få mere at vide
Amberlite 200, MP Biomedicals™
Artikelnummer. 11443111 Shop alle MP Biomedicals produkter
Skift visning
Mængde:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
11443111 500 g
11433111 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11443111 Leverandør MP Biomedicals Leverandørnr. 0215027890
Strongly acidic macroreticular-type cation exchange resin. Sodium form sulfonic acid functionality
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 12626-25-4
MDL nummer MFCD00145784
Tekniske data
Kemisk navn eller materiale Amberlite
Fysisk form Pulver
Farve Gul
Mængde 500 g