Learn More
Fisherbrand™ Amber Wide Mouth Packer Bottles with White Polypropylene Pulp/Vinyl Cap
Ideal general purpose bottle for solids. Fisherbrand™ Amber Wide Mouth Packer Bottles with White Polypropylene Pulp/Vinyl Cap are often used in chemical and environmental applications.
Brand:  Fisherbrand™
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15865472
  • 412.14 DKK / Pack of 84
Estimated Shipment
14-11-2018
Add to basket
Includes: Polypropylene Pulp/Vinyl lined cap
Description
Description
  • Features wide mouth opening for easy transfer of contents
  • Amber color filters out ultraviolet wavelengths, ideal for light sensitive solids
  • Classic “vitamin bottles” and are often used in pharmaceutical packaging
Poplypropylene Pulp/Vinyl Lined Caps
  • Good impact strength and pliability
  • Can be remelted after they are formed
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
Specifications
250mL
Amber, Amber
84/Cs.
White
Wide
Round
Polypropylene
Glass, Glass
Bottle, Wide Mouth Packer, Wide Mouth Packer Bottle
Pulp/Vinyl
45-400
Screw
Videos