Learn More
Fisherbrand™ Custom Cleaned Amber Boston Round Bottles
586.19 DKK valid until 2024-04-30
Use promo code "21749" to get your promotional price.
Use promo code "21749" to get your promotional price.
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Description
- Narrow mouth
- Pre-cleaned for visible particulates using deionised water
- With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Capacity | 2.02 oz., 60mL |
Closure Liner | Polyethylene |
Closure Material | PTFE |
Closure Size | 20-400 mm |
Closure Type | Screw Cap |
Product Type | Boston Round Bottle |
Capacity (Metric) | 60 mL |
Capacity (English) | 2.02 oz. |
Cleaning Procedure | USP purified Water |
Color | Amber |