Learn More
Lenz Laborglasinstrumente™ Amber Borosilicate Glass Separating Funnel with PTFE Plug
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Amber Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into solvent phases of specific densities.
Brand:  Lenz Laborglasinstrumente™ 04008149
Product Code. 11983441
  • 2100.00 DKK / Pack of 2
Only null left
Estimated Shipment: 30-09-2025
Description
  • With NS socket and PE stopper
  • Without scale
  • With PTFE plug
  • With screwthread retaining device
  • Material: borosilicate glass 3.3;
Specifications
Specifications
250 mL
PTFE plug and screw thread retaining device
Separatory Funnel
Borosilicate Glass
29/32 NS Socket
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Amber Borosilicate Glass Separating Funnel with PTFE Plug > Capacity: 250mL