Få mere at vide
Ambenonium Chloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ambenonium Chloride |
| CAS | 115-79-7 |
| Analyt- eller komponentnavne | Ambenonium Chloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C28 H42 Cl2 N4 O2 . 2 Cl |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benzenemethanaminium, N,N'-[(1,2-dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)]bis[2-chloro-N,N-diethyl-, chloride (1:2), Ammonium, [oxalylbis(iminoethylene)]bis[(o-chlorobenzyl)diethyl-, dichloride (8CI), Benzenemethanaminium, N,N'-[(1,2-dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)]bis[2-chloro-N,N-diethyl-, dichloride (9CI), [Oxalylbis(iminoethylene)]bis[(o-chlorobenzyl)diethylammonium chloride] (6CI,7CI), Ambenonium chloride, Ambenonium dichloride, Ambestigmin chloride, Misuran, Mysuran, Mysuran chloride, Mytelase, Mytelase chloride, N,N'-Bis-(2-diethylaminoethyl)oxamide bis-2-chlorobenzylchloride, N,N'-Bis-2-[(2-chlorobenzyl)diethylammonium chloride]-ethyloxamide, Oxamizil, Oxazil, Oxazyl, Win 8077 |
| InChI formel | InChI=1S/C28H40Cl2N4O2.2ClH/c1-5-33(6-2,21-23-13-9-11-15-25(23)29)19-17-31-27(35)28(36)32-18-20-34(7-3,8-4)22-24-14-10-12-16-26(24)30;;/h9-16H,5-8,17-22H2,1-4H3;2*1H |
| Løsningstype | Neat |
| SMIL | [Cl-].[Cl-].CC[N+](CC)(CCNC(=O)C(=O)NCC[N+](CC)(CC)Cc1ccccc1Cl)Cc2ccccc2Cl |
| Vis mere |