Få mere at vide
Ambazone, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ambazone |
| CAS | 539-21-9 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C8 H11 N7 S |
| Mængde | 100 mg |
| Synonym | Hydrazinecarbothioamide, 2-[4-[2-(aminoiminomethyl)hydrazinylidene]-2,5-cyclohexadien-1-ylidene]-Guanidine, [(4-oxo-2,5-cyclohexadien-1-ylidene)amino]-, thiosemicarbazone (6CI,7CI,8CI), Hydrazinecarbothioamide, 2-[4-[(aminoiminomethyl)hydrazono]-2,5-cyclohexadien-1-ylidene]- (9CI), (4-Oxo-2,5-cyclohexadien-1-ylideneamino)guanidine thiosemicarbazone, 1,4-Benzoquinone amidinohydrazone thiosemicarbazone, Ambazon, Ambazone, Anginon, Benzoquinone guanylhydrazone thiosemicarbazone, DC 0572, Faringosept, Guanothiazon, Inversal, Iversal, Ivertol, N-Guanidino-N'-thioureido-p-benzoquinonediimide, Primal, p-Benzoquinone amidinohydrazone thiosemicarbazone |
| InChI formel | InChI=1S/C8H11N7S/c9-7(10)14-12-5-1-3-6(4-2-5)13-15-8(11)16/h1-4H,(H4,9,10,14)(H3,11,15,16) |
| SMIL | NC(=N)NN=C1C=CC(=NNC(=S)N)C=C1 |
| IUPAC navn | [[4-(carbamimidoylhydrazinylidene)cyclohexa-2,5-dien-1-ylidene]amino]thiourea |
| Molekylvægt (g/mol) | 237.28 |
| Vis mere |