Få mere at vide
Tocris Bioscience™ AM 4113
GREENER_CHOICE
Selective, high affinity CB1 antagonist
Brand:  Tocris Bioscience™ 6193/50
Encompass
Produktkode 15746379
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
AM 4113 is a high affinity and selective CB1 antagonist (Kd = 0.89 nM). Exhibits 100-fold selectivity for CB1 over CB2. Attenuates CB1 agonist AM 411-induced locomotor suppression and reduces food intake in vivo, but does not induce signs of nausea.
Tekniske data
Tekniske data
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
614726-85-1
>98%
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner