Få mere at vide
AM 0902, Tocris Bioscience™
GREENER_CHOICE
Potent and selective TRPA1 antagonist
Brand:  Tocris Bioscience 5914/50
Encompass
Produktkode 15973231
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
AM 0902 is a potent and selective TRPA1 antagonist (IC50 = 0.02 μM for human TRPA1). Exhibits no siginificant activity against human TRPV1 or TRPV4, and rat TRPV1, TRPV3 or TRPM8. Exhibits efficacy in a rat pain model. Orally bioavailable.
Tekniske data
Tekniske data
1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-purin-6-one
1883711-97-4
>98%
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner