Få mere at vide
Alverine-d5 Citrate, TRC
Artikelnummer. 30311905
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30311905 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311905 Leverandør Toronto Research Chemicals Leverandørnr. TRCA575782100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alverine-d5 Citrate
CAS 1215327-00-6
Molekylær formel C20 D5 H22 N . C6 H8 O7
Mængde 100 mg
Synonym N-(Ethyl-d5)-N-(3-phenylpropyl)benzenepropanamine 2-Hydroxy-1,2,3- propanetricarboxylate, N-(Ethyl-d5)-3,3'-diphenyldipropylamine Citrate, Antispasmin-d5, Calmabel-d5, Gamatran-d5, NSC 35459-d5, Profenine-d5, Prophelan-d5, Proverine-d5, Spacolin-d5, Spasmaverine-d5, Spasmonal-d5
InChI formel InChI=1S/C20H27N.C6H8O7/c1-2-21(17-9-15-19-11-5-3-6-12-19)18-10-16-20-13-7-4-8-14-20;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-8,11-14H,2,9-10,15-18H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1D3,2D2;
SMIL [2H]C([2H])([2H])C([2H])([2H])N(CCCc1ccccc1)CCCc2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC navn 2-hydroxypropane-1,2,3-tricarboxylic acid;N-(1,1,2,2,2-pentadeuterioethyl)-3-phenyl-N-(3-phenylpropyl)propan-1-amine
Molekylvægt (g/mol) 478.589
Formel vægt 478.273