missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hexadecahydrate, Honeywell

Brand:  Honeywell Research Chemicals 06421-250G

Additional Details : CAS Number : 16828-11-8 Weight : 0.25000kg

Quantity 250g

 View more versions of this product

Product Code. 15640830

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance
View alternative products »


Specifications

Chemical Identifiers

16828-11-8
≥95% (KT)
purum p.a.
MFCD00149137
Aluminum Sulfate Hexadecahydrate
630.4g/mol
Al2(SO4)3 · 16H2O
3260

Specifications

Aluminum Sulfate Hexadecahydrate
purum p.a.
Al2(SO4)3 · 16H2O
MFCD00149137
250g
16828-11-8
≥95% (KT)
630.4g/mol
3260
Videos
SDS