missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Solution for ICP, 1g/L HCl matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical S002250

Additional Details : Weight : 0.25000kg

Quantity 250mL

 View more versions of this product

Product Code. 12943354

  • 482.00 DKK / 250mL

Estimated Shipment
07-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

Specifications

1g/L
HCl
250mL
Videos
SDS
Documents