Få mere at vide
Aluminum Phthalocyanine Chloride (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Aluminum Phthalocyanine Chloride (Technical Grade) |
| CAS | 14154-42-8 |
| Molekylær formel | C32H16AlClN8 |
| Mængde | 10 mg |
| Synonym | (SP-5-12)-chloro[29H,31H-phthalocyaninato(2-)-N29,N30,N31,N32]-Aluminum, chloro[phthalocyaninato(2-)]-Aluminum (6CI,7CI,8CI), (SP-5-12)-Chloro[29H,31H-phthalocyaninato(2-)-kN29,kN30,kN31,kN32]aluminum, aluminum 29H,31H-Phthalocyanine complex, C.I. 741300, C.I. Pigment Blue 79, Chloro(phthalocyaninato)aluminum, Chloro[phthalocyaninato(2-)]aluminum, Chlorophthalocyaninealuminum, Monochloroaluminum phthalocyanine, Pigment Blue 79, SF Cyan A 209 |
| InChI formel | InChI=1S/C32H16N8.Al.ClH/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;;1H/q-2;+3;/p-1 |
| SMIL | [Cl-][Al+3]123N4=C5N=C6c7ccccc7C(=NC8=N1C(=NC9=C%10C=CC=CC%10=C(N=C4c%11ccccc5%11)[N-]29)c%12ccccc8%12)[N-]36 |
| Molekylvægt (g/mol) | 574.97 |
| Formel vægt | 574.1002 |