Få mere at vide
Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, M Plane, Thermo Scientific™
Mængde:
1 Ea.
5 Ea.
Pakningsstørrelse:
Hver
Pakke med 5
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Aluminum Oxide Substrate, M Plane |
| Navn note | Polished One Side |
| Mængde | 1 Ea. |
| Molekylær formel | Al2O3 |
| MDL nummer | MFCD00003424 |