missing translation for 'onlineSavingsMsg'
Learn More
Aluminum oxide, single crystal, Thermo Scientific™ Product Code.: 15481627

Aluminum oxide, single crystal, Thermo Scientific™

Product Code. 15481627
5 Pc., Pack of 5
Click to view available options
Quantity:
1 Pc.
5 Pc.
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 15481627

Brand: Thermo Scientific Alfa Aesar 040257.KJ

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a grinding or blending agent. It is essential in protein extraction as a component in the preparation of tissues. It is a source of aluminum in reactions, an abrasive agent and as a refractory material.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Quantity 5 Pc.
Solubility Information Soluble in water (0.0098g/L at 29°C),hexane and toluene.
IUPAC Name dialuminium(3+) trioxidandiide
Packaging Shrink-wrapped
Chemical Name or Material Aluminum oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum oxide, single crystal, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.