missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, single crystal, Thermo Scientific™

Catalog Number p-7076381
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]

Used as a grinding or blending agent. It is essential in protein extraction as a component in the preparation of tissues. It is a source of aluminum in reactions, an abrasive agent and as a refractory material.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 Pc.
Solubility Information Soluble in water (0.0098g/L at 29°C),hexane and toluene.
IUPAC Name dialuminum;oxygen(2-)
Packaging Shrink-wrapped
Chemical Name or Material Aluminum oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'