missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, Puriss., ≥98% Al2O3 basis, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 500 g |
Linear Formula | Al2O3 |
UN Number | NONH for all modes of transport |
Formula Weight | 101.96g/mol |
Percent Purity | ≥98% (Al2O3 basis) |
Grade | puriss. |
Chemical Name or Material | Aluminum Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'