missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Aluminum oxide, neutral, for TLC, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Boiling Point | 2980°C |
Melting Point | 2045°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | Al2O3 |
Merck Index | 14,356 |
Solubility Information | Slightly soluble in alkaline solutions. Insoluble in water. |
Formula Weight | 101.96 |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'