missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Activity Grade Super I, Type WN-6, Honeywell Fluka™

Catalog Number p-7120148
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]

Type WN-6, Neutral, Activity Grade Super I

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 2980°C
Melting Point 2072°C
Color Colorless
Quantity 100 g
Linear Formula Al2O3
UN Number NONH for all modes of transport
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96g/mol
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 3960g/cm3 at 20°C
Chemical Name or Material Aluminum oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'