missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum foil, 1.0mm (0.04 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%

Brand:  Alfa Aesar™ 014192.VK

Additional Details : CAS Number : 7429-90-5 Weight : 0.01000kg

Quantity 5 x 45 x 240cm

 View more versions of this product

Product Code. 15484755

  • 7047.77 DKK / Each


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
CHEBI:33629
[Al]

Specifications

Aluminum foil
5 x 45 x 240cm
7429-90-5
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99%
≈729g/45x60cm
1.0mm (0.04 in.) thick, Annealed, Temper: soft
Si and Fe (combined) typically 1%
Al
aluminium, metal, powder, aluminio, metana, adom, alumina fibre, aluminium flake, dust, noral aluminium
[Al]
Odorless
5359268
Foil
Videos
SDS Documents