Learn More
Bochem™ Aluminum Bossheads
Product Code. 10666951
Change view
Click to view available options
Diameter (Metric) Outer:
16.2 mm
16.5 mm
27 mm
Thread Size:
8
10
Weight (Metric):
0.06 kg
0.09 kg
0.13 kg
0.3 kg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Thread Size Weight (Metric) Unit Size
10666951 27 mm 10 0.3 kg Each
10189590 16.2 mm 8 0.06 kg Each
10751392 16.5 mm 8 0.13 kg Each
10202311 16.2 mm 8 0.09 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10666951 Supplier Bochem™ Supplier No. 5352
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Forventet forsendelse: 22-12-2026
Manufactured from aluminum. Bochem™ Aluminum Bossheads are DIN 12895 certified.
  • Manufactured from aluminum
  • DIN 12895
  • Type 1: Thumb screw M8
Specifications
Material Aluminum
Diameter (Metric) Outer 27 mm
Product Type Aluminum Bossheads
Thread Size 10
Quantity 1 Ea.
Weight (Metric) 0.3 kg