missing translation for 'onlineSavingsMsg'
Learn More

Altrenogest, TRC

Product Code. 18082310
Change view
Click to view available options
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18082310 100 mg
17919839 50 mg
18033310 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18082310 Supplier Toronto Research Chemicals Supplier No. TRCA575755100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850-52-2
Molekylær formel C21 H26 O2
Molekylvægt (g/mol) 310.43
Synonym Estra-4,9,11-trien-3-one, 17-hydroxy-17-(2-propenyl)-, (17β)- (9CI), Estra-4,9,11-trien-3-one, 17α-allyl-17-hydroxy- (7CI,8CI), (17β)-17-Hydroxy-17-(2-propen-1-yl)estra-4,9,11-trien-3-one, A 35957, Allyltrenbolone, Altrenogest, DRC 6246, Matrix, Matrix (progestogen), R 2267, RU 2267, Regumate
IUPAC navn (8S,13S,14S,17R)-17-hydroxy-13-methyl-17-prop-2-enyl-1,2,6,7,8,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMIL C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@]2(O)CC=C

Specifications

Kemisk navn eller materiale Altrenogest
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C21H26O2/c1-3-10-21(23)12-9-19-18-6-4-14-13-15(22)5-7-16(14)17(18)8-11-20(19,21)2/h3,8,11,13,18-19,23H,1,4-7,9-10,12H2,2H3/t18-,19+,20+,21+/m1/s1
Formel vægt 310.1933
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.