missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ ALS Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16618454
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16618454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16618454 Leverandør Invitrogen™ Leverandørnr. PA5101977

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IGFALS.

The protein encoded by this gene is a serum protein that binds insulin-like growth factors, increasing their half-life and their vascular localization. Production of the encoded protein, which contains twenty leucine-rich repeats, is stimulated by growth hormone. Defects in this gene are a cause of acid-labile subunit deficiency, which maifests itself in a delayed and slow puberty. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ALS
Applikationer Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGFALS
Genadgangsnr. P35858
Gene Alias ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111
Gensymboler IGFALS
Værtsarter Rabbit
Immunogen A synthesized peptide derived from human IGFALS(Accession P35858), corresponding to amino acid residues S136-P186.
Oprensningsmetode Affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 3483
Målarter Human
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.