Få mere at vide
Alprenolol Hydrochloride, TRC
Artikelnummer. 30134006
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134006 50 mg
17901875 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134006 Leverandør Toronto Research Chemicals Leverandørnr. TRCA57568550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13707-88-5
Molekylær formel C15 H23 N O2 . Cl H
Synonym Alprenolol hydrochloride∣(2RS)-1-[(1-Methylethyl)amino]-3-[2-(prop-2-enyl)phenoxy]propan-2-ol hydrochloride∣2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yl)phenoxy]-, hydrochloride (1:1)∣2-Propanol, 1-(o-allylphenoxy)-3-(isopropylamino)-, hydrochloride (8CI)∣2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyl)phenoxy]-, hydrochloride (9CI)∣(±)-1-(o-Allylphenoxy)-3-isopropylamino-2-propanol hydrochloride∣(±)-Alprenolol hydrochloride∣1-(2-Allylphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride∣1-(2-Allylphenoxy)-3-(isopropylamino)propan-2-ol hydrochloride∣1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride∣Alfeprol∣Apllobal∣Applobal∣Aprobal∣Aptin∣Aptine∣Aptol Duriles∣Betaptin∣DL-1-(o-Allylphenoxy)-3-isopropylamino-2-propanol hydrochloride∣Dimacor∣Gubernal∣H 56/28∣Regletin∣Yobir∣[2-Hydroxy-3-[2-(prop-2-en-1-yl)phenoxy]propyl](propan-2-yl)amine hydrochloride∣dl-Alprenolol chloride
Tekniske data
Kemisk navn eller materiale Alprenolol hydrochloride
Mængde 50 mg
Forsendelsestilstand Room Temperature
Formel vægt 285.1496
Molekylvægt (g/mol) 285.81
InChI nøgle RRCPAXJDDNWJBI-UHFFFAOYNA-N
SMIL [H+].[Cl-].CC(C)NCC(O)COC1=CC=CC=C1CC=C
IUPAC navn hydrogen 1-[2-(prop-2-en-1-yl)phenoxy]-3-[(propan-2-yl)amino]propan-2-ol chloride