Få mere at vide
Alprenolol hydrochloride, Dr. Ehrenstorfer
Artikelnummer. 30320232
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30320232 10 mg
1 options
Artikelnummer. 30320232 Leverandør Dr. Ehrenstorfer Leverandørnr. DRECA10142650
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alprenolol hydrochloride
CAS 13707-88-5
Analyt- eller komponentnavne Alprenolol hydrochloride
Forsendelsestilstand +20°C
Molekylær formel C15 H23 N O2 . Cl H
Certificeringer/compliance ISO/IEC 17025
Synonym Alprenolol hydrochloride, (2RS)-1-[(1-Methylethyl)amino]-3-[2-(prop-2-enyl)phenoxy]propan-2-ol hydrochloride, 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propen-1-yl)phenoxy]-, hydrochloride (1:1), 2-Propanol, 1-(o-allylphenoxy)-3-(isopropylamino)-, hydrochloride (8CI), 2-Propanol, 1-[(1-methylethyl)amino]-3-[2-(2-propenyl)phenoxy]-, hydrochloride (9CI), (±)-1-(o-Allylphenoxy)-3-isopropylamino-2-propanol hydrochloride, (±)-Alprenolol hydrochloride, 1-(2-Allylphenoxy)-2-hydroxy-3-isopropylaminopropane hydrochloride, 1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride, Alfeprol, Alprenolol hydrochloride, Apllobal, Applobal, Aprobal, Aptin, Aptine, Aptol Duriles, Betaptin, DL-1-(o-Allylphenoxy)-3-isopropylamino-2-propanol hydrochloride, Dimacor, Gubernal, H 56/28, Regletin, Yobir, dl-Alprenolol chloride
InChI formel InChI=1S/C15H23NO2.ClH/c1-4-7-13-8-5-6-9-15(13)18-11-14(17)10-16-12(2)3;/h4-6,8-9,12,14,16-17H,1,7,10-11H2,2-3H3;1H
SMIL Cl.CC(C)NCC(O)COc1ccccc1CC=C
IUPAC navn 1-(propan-2-ylamino)-3-(2-prop-2-enylphenoxy)propan-2-ol;hydrochloride
Molekylvægt (g/mol) 285.81
Mængde 10 mg
Formel vægt 285.1496 g/mol