Få mere at vide
Alpidem-d14, TRC
Artikelnummer. 30309405
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30309405 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309405 Leverandør Toronto Research Chemicals Leverandørnr. TRCA57556210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alpidem-d14
CAS 1189962-23-9
Molekylær formel C21H9D14Cl2N3O
Mængde 10 mg
Synonym 6-Chloro-2-(4-chlorophenyl)-N,N-(dipropyl-14)-imidazo[1,2-a]pyridine-3-acetamide, SL 80.0342-00-d14
InChI formel InChI=1S/C21H23Cl2N3O/c1-3-11-25(12-4-2)20(27)13-18-21(15-5-7-16(22)8-6-15)24-19-10-9-17(23)14-26(18)19/h5-10,14H,3-4,11-13H2,1-2H3/i1D3,2D3,3D2,4D2,11D2,12D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])N(C(=O)Cc1c(nc2ccc(Cl)cn12)c3ccc(Cl)cc3)C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 2-[6-chloro-2-(4-chlorophenyl)imidazo[1,2-a]pyridin-3-yl]-N,N-bis(1,1,2,2,3,3,3-heptadeuteriopropyl)acetamide
Molekylvægt (g/mol) 418.42