Få mere at vide
Alphaxalone-d5 (major), TRC
Artikelnummer. 30326720
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30326720 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326720 Leverandør Toronto Research Chemicals Leverandørnr. TRCA57553210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Alphaxalone-d5
CAS 83014-68-0
Molekylær formel C21H27D5O3
Mængde 10 mg
Synonym 3α-Hydroxy-5α-pregnane-11,20-dione-d5, 5α-Alphaxalone-d5, 5α-Pregnan-3α-ol-11,20-dione-d5, 5α-Pregnane-3α-ol-11,20-dione-d5, Alfadione-d5, Alfaxalone-d5, GR 2/234-d5
InChI formel InChI=1S/C21H32O3/c1-12(22)16-6-7-17-15-5-4-13-10-14(23)8-9-20(13,2)19(15)18(24)11-21(16,17)3/h13-17,19,23H,4-11H2,1-3H3/t13-,14+,15-,16+,17-,19+,20-,21+/m0/s1/i8D2,10D2,14D
SMIL O[C@](C1([2H])[2H])([2H])C([2H])([2H])C[C@@]2(C)[C@@]1([H])CC[C@]3([H])[C@]2([H])C(C[C@@]4(C)[C@@]3([H])CC[C@@H]4C(C)=O)=O
IUPAC navn (3R,5S,8S,9S,10S,13S,14S,17S)-17-acetyl-2,2,3,4,4-pentadeuterio-3-hydroxy-10,13-dimethyl-5,6,7,8,9,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-11-one
Molekylvægt (g/mol) 337.51