Få mere at vide
Ansell™ AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and Taped
Shop alle Ansell produkterBeskrivelse
- The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and taped - Model 214 chemical protective suit is made with a multi-layer barrier fabric, which offers wearers excellent protection against several chemicals, including inorganic and biological hazards
- These highly visible, bright yellow chemical protection coveralls improve worker safety in dimly lit environments
- Featuring welded and taped seams, this full chemical protection suit offers a strong barrier against harmful liquid and particles
Tekniske data
Tekniske data
| Type | Apron |
| Størrelse | Small |
| Materiale | Multi-layer Barrier Laminate Fabric |
| Farve | Yellow |
| Engangs | Single-use |
| Lukketype | Hook and Loop (Rear) |
| Sterilitet | Non-sterile |
| Latex fri | Yes |
| Antal lommer | None |
| Sømtype | Welded and Taped |
| Vis mere |
Product Title
Individuate un'opportunità di miglioramento?