Learn More
Ansell AlphaTec™ 2000 Ts PLUS
Chemical protective suits that have a proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards.
Brand: Ansell WH20T-00103-06
3430.36 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Specifications
2X-Large | |
White | |
EN 374:2003, , EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003, DIN 32781:2010 | |
Body Painting Including Surface Inspection, Cleaning of Plant and Machines, Fibre Glass/Resin Applications/Ceramic Fibres, Product Inspection, Loading and Hauling | |
W20T103 |
Microporous Non-woven Polyethylene Laminate | |
Stitched, Taped | |
Coverall with Collar | |
63 gsm |
Product Content Correction
Product Title
Ansell AlphaTec™ 2000 Ts PLUS > Size: 2X-Large, Style: W20T103
Spot an opportunity for improvement?Share a Content Correction