Learn More
Ansell AlphaTec™ 2000 Lab Coats, Model 209
Chemical resistant lab Coat features breathable microporous laminate technology.
2160.00 DKK - 4075.00 DKK
Specifications
Product Type | Chemical Resistant Lab Coat |
---|---|
Gender | Unisex |
Material | Microporous Polyethylene Laminate |
Color | White |
Closure Type | Zipper |
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Chemical Resistant Lab Coat | |
Microporous Polyethylene Laminate | |
Zipper | |
Limited-use | |
Chemicals | |
2 |
Unisex | |
White | |
Elastic | |
Non-sterile | |
Standard Length | |
CE Category III, EN 14126 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell AlphaTec™ 2000 Lab Coats, Model 209