Få mere at vide
Skytec™ AlphaShield - S2200 Disposable Coverall
Størrelse:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Lightweight microporous polyethylene (PE) laminate
- Bound seam for additional protection and strength
- 2-way front zipper with re-sealable storm flap
- Thumb loop ensures arm protection remains in place when arm is raised
- Elasticated waist to enhance fit
- Innovative three-piece elasticated hood construction provides added comfort, durability and fit
- Elasticated ankles and wrists
Specifications
Specifications
| Produkttype | Chemical-resistant Coveralls |
| Størrelse | Medium |
| Køn | Unisex |
| Materiale | Spunbond Polypropylene |
| Farve | White |
| Engangs | Single-use |
| Talje stil | Elastic |
| Ankel stil | Elastic |
| Manchet stil | Elastic |
| Hals stil | Hooded |
| Show More |