Få mere at vide
(AlphaS,BetaR)-Bedaquiline, TRC
Artikelnummer. 30133344
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133344 25 mg
30138533 1 mg
17974425 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133344 Leverandør Toronto Research Chemicals Leverandørnr. TRCB11954025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 843663-66-1
Molekylær formel C32 H31 Br N2 O2
Molekylvægt (g/mol) 555.5
Synonym 3-Quinolineethanol, 6-bromo-α-[2-(dimethylamino)ethyl]-2-methoxy-α-1-naphthalenyl-β-phenyl-, (αS,βR)-, (αS,βR)-6-Bromo-α-[2-(dimethylamino)ethyl]-2-methoxy-α-1-naphthalenyl-β-phenyl-3-quinolineethanol, Bedaquiline, R 207910, Sirturo, TMC 207, (αS,βR)-Bedaquiline
IUPAC navn (1R,2S)-1-(6-bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-naphthalen-1-yl-1-phenylbutan-2-ol
SMIL COc1nc2ccc(Br)cc2cc1[C@@H](c3ccccc3)[C@@](O)(CCN(C)C)c4cccc5ccccc45
Tekniske data
Kemisk navn eller materiale Bedaquiline
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C32H31BrN2O2/c1-35(2)19-18-32(36,28-15-9-13-22-10-7-8-14-26(22)28)30(23-11-5-4-6-12-23)27-21-24-20-25(33)16-17-29(24)34-31(27)37-3/h4-17,20-21,30,36H,18-19H2,1-3H3/t30-,32-/m1/s1
Formel vægt 554.1569
Procent renhed >95%