Få mere at vide
(AlphaR,BetaS)-rel-Bedaquiline-d6, TRC
Artikelnummer. 30343024
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30343024 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343024 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1195575MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (aR,bS)-rel-Bedaquiline-d6
CAS 2271264-27-6
Molekylær formel C32H25D6BrN2O2
Mængde 5 mg
Synonym (αR,βS)-rel-6-Bromo-α-[2-(dimethylamino)ethyl]-2-methoxy-α-1-naphthalenyl-β-phenyl-3-quinolineethanol-d6
InChI formel InChI=1S/C32H31BrN2O2/c1-35(2)19-18-32(36,28-15-9-13-22-10-7-8-14-26(22)28)30(23-11-5-4-6-12-23)27-21-24-20-25(33)16-17-29(24)34-31(27)37-3/h4-17,20-21,30,36H,18-19H2,1-3H3/t30?,32-/m1/s1/i1D3,2D3
SMIL BrC1=CC=C2C(C=C([C@@H](C3=CC=CC=C3)[C@@](O)(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])C4=C(C=CC=C5)C5=CC=C4)C(OC)=N2)=C1
IUPAC navn (1R,2S)-4-(bis(methyl-d3)amino)-1-(6-bromo-2-methoxyquinolin-3-yl)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol
Molekylvægt (g/mol) 561.54
Formel vægt 560.19