missing translation for 'onlineSavingsMsg'
Få mere at vide

(alphaR)-(1S,2S,3R,5S)-Pinanediol-1-amino-3-methylbutane-1-boronate Trifluoroacetic Acid Salt, TRC

Artikelnummer. 30427579
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30427579 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427579 Supplier Toronto Research Chemicals Supplier No. TRCP4585051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (1R)-(S)-Pinanediol-1-amino-3-methylbutane-1-boronate trifluoroacetate salt
CAS 179324-87-9
Molekylær formel C15 H28 B N O2 . C2 H F3 O2
Mængde 1 g
Synonym 4,6-Methano-1,3,2-benzodioxaborole-2-methanamine','hexahydro-3a,8,8-trimethyl-α-(2-methylpropyl)-','(αR,3aS,4S,6S,7aR)-','2,2,2-trifluoroacetate (1:1), 4,6-Methano-1,3,2-benzodioxaborole-2-methanamine','hexahydro-3a,5,5-trimethyl-α-(2-methylpropyl)-','(αR,3aS,4S,6S,7aR)-','trifluoroacetate (9CI), 4,6-Methano-1,3,2-benzodioxaborole-2-methanamine','hexahydro-3a,5,5-trimethyl-α-(2-methylpropyl)-','[3aS-[2(S*),3aα,4β,6β,7aα]]-','trifluoroacetate, (R)-BoroLeu-(+)-Pinanediol Trifluoroacetate
InChI formel InChI=1S/C15H28BNO2.C2HF3O2/c1-9(2)6-13(17)16-18-12-8-10-7-11(14(10,3)4)15(12,5)19-16;3-2(4,5)1(6)7/h9-13H,6-8,17H2,1-5H3;(H,6,7)/t10-,11-,12+,13-,15-;/m0./s1
SMIL CC(C)C[C@H](N)B1O[C@@H]2C[C@@H]3C[C@@H](C3(C)C)[C@]2(C)O1.OC(=O)C(F)(F)F
Molekylvægt (g/mol) 379.22
Formel vægt 379.2142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.