Beskrivelse
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human alpha Tubulin The alpha Tubulin Recombinant Protein Antigen is derived from E. coli. The alpha Tubulin Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Tekniske data
Tekniske data
| Protein længde | ATYAPVISAEKAYHEQLSVAEITN |
| Renhed | >80% by SDS-PAGE and Coomassie blue staining |
| Fælles navn | TUBA1A Recombinant Protein Antigen |
| Indhold og opbevaring | Store at −20°C. Avoid freeze-thaw cycles. |
| Formulering | PBS and 1M Urea, pH 7.4 |
| Til brug med (applikation) | AC |
| Gene Alias | Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific |
| Gen symbol | TUBA1A |
| Etikettype | Unlabeled |
| Molekylvægt (g/mol) | 20 kDa |
| Vis mere |
For Research Use Only.