missing translation for 'onlineSavingsMsg'
Få mere at vide

alpha-Tocopherol Phosphate Disodium Salt, TRC

Artikelnummer. 30449010
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30449010 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30449010 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5261855MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale alpha-Tocopherol Phosphate Disodium Salt
CAS 90940-45-7
Molekylær formel C29 H49 Na2 O5 P
Mængde 5 mg
Synonym α-Tocopherol Phosphate Disodium Salt
InChI formel InChI=1S/C29H51O5P.2Na/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(34-35(30,31)32)23(5)24(6)28(26)33-29;;/h20-22H,9-19H2,1-8H3,(H2,30,31,32);;/q;2*+1/p-2/t21-,22-,29-;;/m1../s1
SMIL CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OP(=O)(O[Na])O[Na])c(C)c(C)c2O1
IUPAC navn [sodiooxy-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]phosphoryl]oxysodium
Molekylvægt (g/mol) 554.65
Formel vægt 554.311
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.