Få mere at vide
alpha-Tocopherol-d6, TRC
Artikelnummer. 30449772
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30449772 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449772 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5261271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-Tocopherol-d6
CAS 113892-08-3
Renhedsgrad noter HPLC
Molekylær formel C29 2H6 H44 O2
Mængde 1 mg
Synonym all-rac-alpha-Tocopherol D6 (5,7-Dimethyl D6)
InChI formel InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/i5D3,7D3
SMIL [2H]C([2H])([2H])c1c(C)c2OC(C)(CCCC(C)CCCC(C)CCCC(C)C)CCc2c(c1O)C([2H])([2H])[2H]
IUPAC navn 2,8-dimethyl-5,7-bis(trideuteriomethyl)-2-(4,8,12-trimethyltridecyl)-3,4-dihydrochromen-6-ol
Molekylvægt (g/mol) 436.74
Formel vægt 436.4187
Procent renhed >95