Få mere at vide
Alpha-Thujene, TRC
Artikelnummer. 17992925
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17992925 5 mg
17924205 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17992925 Leverandør Toronto Research Chemicals Leverandørnr. TRCT4058005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2867-05-2
Molekylær formel C10H16
Molekylvægt (g/mol) 136.23
Synonym 3-Thujene, 2-Methyl-5-(1-methylethyl)bicyclo[3.1.0]hex-2-ene, (±)-α-Thujene, 2-Methyl-5-isopropylbicyclo[3.1.0]-2-hexene, Origanene, 2-Methyl-5-(1-methylethyl)-bicyclo[3.1.0]hex-2-ene
IUPAC navn rac-(1R)-2-methyl-5-propan-2-ylbicyclo[3.1.0]hex-2-ene
SMIL CC(C)[C@@]12CC=C(C)[C@@H]1C2
Tekniske data
Kemisk navn eller materiale Alpha-Thujene
Mængde 5 mg
InChI formel InChI=1S/C10H16/c1-7(2)10-5-4-8(3)9(10)6-10/h4,7,9H,5-6H2,1-3H3/t9-,10+/m0/s1
Formel vægt 136.1252