Få mere at vide
Alpha-Terpineol-d3 (Propyl Methyl-d3), TRC
Artikelnummer. 30419818
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30419818 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419818 Leverandør Toronto Research Chemicals Leverandørnr. TRCT11065250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-Terpineol alpha-methyl-d3
CAS 203633-12-9
Molekylær formel C10 2H3 H15 O
Mængde 50 mg
Synonym 3-Cyclohexene-1-methanol','α,4-dimethyl-α-(methyl-d3)- (9CI), α-Terpineol-d3, α-Terpineol-D3 (α-methyl D3)
InChI formel InChI=1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3/i2D3
SMIL [2H]C([2H])([2H])C(C)(O)C1CCC(=CC1)C
IUPAC navn 1,1,1-trideuterio-2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Molekylvægt (g/mol) 157.27
Formel vægt 157.1546